Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GTF2B
Entrez Gene ID
2959
Gene Name
general transcription factor IIB
Gene Aliases
TF2B, TFIIB
Chromosome Location
Chr.1: 88852638 - 88891944 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001514.5 NP_001505.1 1 - 2 136 66
GenBank mRNA AK289822.1 - 1 - 2 82 66
BC020597.1 - 1 - 2 82 66
BP379342.1 - 1 - 2 136 66
M76766.1 - 1 - 2 55 66

View Full Product Details