Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAP4
Entrez Gene ID
4134
Gene Name
microtubule associated protein 4
Gene Aliases
-
Chromosome Location
Chr.3: 47850690 - 48089279 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134364.1 NP_001127836.1 16 - 17 3679 88
NM_002375.4 NP_002366.2 16 - 17 3679 88
XM_005265133.4 XP_005265190.1 19 - 20 6745 88
XM_005265134.4 XP_005265191.1 19 - 20 7158 88
XM_005265155.4 XP_005265212.1 15 - 16 3603 88
XM_005265157.4 XP_005265214.1 9 - 10 2972 88
XM_005265158.2 XP_005265215.1 7 - 8 2765 88
XM_006713146.3 XP_006713209.1 20 - 21 6777 88
XM_006713147.3 XP_006713210.1 20 - 21 7194 88
XM_006713152.3 XP_006713215.1 19 - 20 7143 88
XM_011533703.2 XP_011532005.1 19 - 20 6694 88
XM_011533704.2 XP_011532006.1 19 - 20 6649 88
XM_011533705.1 XP_011532007.1 18 - 19 6643 88
XM_011533708.2 XP_011532010.1 18 - 19 6598 88
XM_011533709.2 XP_011532011.1 18 - 19 6580 88
XM_011533710.2 XP_011532012.1 18 - 19 6571 88
XM_011533718.1 XP_011532020.1 16 - 17 6436 88
XM_011533719.2 XP_011532021.1 17 - 18 3259 88
XM_017006391.1 XP_016861880.1 19 - 20 6649 88
XM_017006392.1 XP_016861881.1 18 - 19 6580 88
XM_017006393.1 XP_016861882.1 19 - 20 6726 88
XM_017006394.1 XP_016861883.1 18 - 19 7020 88
XM_017006395.1 XP_016861884.1 17 - 18 3259 88
XM_017006396.1 XP_016861885.1 17 - 18 3214 88
XM_017006397.1 XP_016861886.1 17 - 18 3214 88
XM_017006398.1 XP_016861887.1 16 - 17 3208 88
XM_017006399.1 XP_016861888.1 17 - 18 3723 88
XM_017006400.1 XP_016861889.1 17 - 18 3723 88
XM_017006401.1 XP_016861890.1 15 - 16 3100 88
XM_017006402.1 XP_016861891.1 16 - 17 3672 88
XM_017006403.1 XP_016861892.1 15 - 16 3052 88
XM_017006404.1 XP_016861893.1 15 - 16 3052 88
XM_017006405.1 XP_016861894.1 16 - 17 3627 88
XM_017006406.1 XP_016861895.1 16 - 17 3609 88
XM_017006407.1 XP_016861896.1 15 - 16 3007 88
XM_017006408.1 XP_016861897.1 15 - 16 3007 88
XM_017006409.1 XP_016861898.1 15 - 16 3558 88
XM_017006410.1 XP_016861899.1 16 - 17 3555 88
XM_017006411.1 XP_016861900.1 15 - 16 3549 88
XM_017006412.1 XP_016861901.1 15 - 16 3549 88
XM_017006413.1 XP_016861902.1 14 - 15 3465 88
XM_017006414.1 XP_016861903.1 14 - 15 3465 88
XM_017006415.1 XP_016861904.1 14 - 15 3420 88
XM_017006416.1 XP_016861905.1 12 - 13 2842 88
XM_017006417.1 XP_016861906.1 9 - 10 2972 88
XM_017006418.1 XP_016861907.1 9 - 10 2927 88
XM_017006419.1 XP_016861908.1 8 - 9 2858 88
XM_017006420.1 XP_016861909.1 8 - 9 2858 88
XM_017006421.1 XP_016861910.1 7 - 8 2765 88
XM_017006422.1 XP_016861911.1 7 - 8 2720 88
GenBank mRNA AB209377.1 - 16 - 17 3239 88
AK299854.1 - 7 - 8 1106 88
AL831878.1 - 3 - 4 258 88
BC008715.2 - 14 - 15 2724 88
BC012794.2 - 14 - 15 2724 88
BC015149.2 - 14 - 15 2724 88
BC048200.1 - 9 - 10 2270 88
CR749544.1 - 7 - 8 1177 88
M64571.1 - 16 - 17 3243 88
U19727.1 - 16 - 17 3228 88

View Full Product Details