The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002431.3 | NP_002422.1 | 5 - 6 | 686 | 94 |
XM_005267687.2 | XP_005267744.1 | 5 - 6 | 692 | 94 | |
XM_005267688.2 | XP_005267745.1 | 5 - 6 | 692 | 94 | |
XM_017021332.1 | XP_016876821.1 | 6 - 7 | 534 | 94 | |
XM_017021333.1 | XP_016876822.1 | 6 - 7 | 825 | 94 | |
XM_017021334.1 | XP_016876823.1 | 3 - 4 | 877 | 94 | |
GenBank mRNA | BC000820.1 | - | 5 - 6 | 665 | 94 |
CR450336.1 | - | 5 - 6 | 564 | 94 | |
KU178191.1 | - | 5 - 6 | 564 | 94 | |
X87843.1 | - | 5 - 6 | 613 | 94 | |
X92669.1 | - | 5 - 6 | 598 | 94 |