The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000256.3 | NP_000247.2 | 12 - 13 | 1146 | 56 |
GenBank mRNA | AK291569.1 | - | 12 - 13 | 1146 | 56 |
AK297843.1 | - | 12 - 13 | 1146 | 56 | |
BC136543.1 | - | 12 - 13 | 1123 | 56 | |
BC136546.1 | - | 12 - 13 | 1123 | 56 | |
BC142685.1 | - | 12 - 13 | 1126 | 56 | |
BC151211.1 | - | 12 - 13 | 1129 | 56 | |
EF560722.1 | - | 12 - 13 | 1147 | 56 |