The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321809.1 | NP_001308738.1 | 4 - 5 | 387 | 103 |
NM_001321810.1 | NP_001308739.1 | 4 - 5 | 387 | 103 | |
NM_001321812.1 | NP_001308741.1 | 3 - 4 | 313 | 103 | |
NM_001321814.1 | NP_001308743.1 | 4 - 5 | 387 | 103 | |
NM_001321815.1 | NP_001308744.1 | 3 - 4 | 262 | 103 | |
NM_001321818.1 | NP_001308747.1 | 4 - 5 | 387 | 103 | |
NM_001321819.1 | NP_001308748.1 | 4 - 5 | 387 | 103 | |
NM_001321821.1 | NP_001308750.1 | 4 - 5 | 387 | 103 | |
NM_002877.5 | NP_002868.1 | 4 - 5 | 387 | 103 | |
NM_133509.3 | NP_598193.2 | 4 - 5 | 387 | 103 | |
NM_133510.3 | NP_598194.1 | 4 - 5 | 387 | 103 | |
XM_011537050.2 | XP_011535352.1 | 4 - 5 | 387 | 103 | |
XM_011537051.2 | XP_011535353.1 | 4 - 5 | 387 | 103 | |
XM_017021545.1 | XP_016877034.1 | 4 - 5 | 387 | 103 | |
GenBank mRNA | AK309787.1 | - | 4 - 5 | 387 | 103 |
AK310686.1 | - | 4 - 5 | 354 | 103 | |
AL597484.1 | - | 4 - 5 | 387 | 103 | |
BC030219.1 | - | 4 - 5 | 345 | 103 | |
BX248061.1 | - | 4 - 5 | 337 | 103 | |
BX248766.1 | - | 4 - 5 | 342 | 103 | |
CR536560.1 | - | 3 - 4 | 310 | 103 | |
U84138.1 | - | 4 - 5 | 364 | 103 | |
U92074.1 | - | 4 - 5 | 380 | 103 | |
Y15571.1 | - | 4 - 5 | 358 | 103 |