The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135669.1 | NP_001129141.1 | 12 - 13 | 1804 | 68 |
NM_004736.3 | NP_004727.2 | 13 - 14 | 1999 | 68 | |
GenBank mRNA | AF089744.1 | - | 13 - 14 | 1972 | 68 |
AF099082.1 | - | 13 - 14 | 1976 | 68 | |
AF115389.1 | - | 13 - 14 | 1807 | 68 | |
AK074789.1 | - | 3 - 4 | 386 | 68 | |
AL133058.1 | - | 10 - 11 | 1501 | 68 | |
AL137583.1 | - | 6 - 7 | 972 | 68 | |
BC041142.1 | - | 12 - 13 | 1782 | 68 |