Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPINT2
Entrez Gene ID
10653
Gene Name
serine peptidase inhibitor, Kunitz type 2
Gene Aliases
DIAR3, HAI-2, HAI2, Kop, PB
Chromosome Location
Chr.19: 38264458 - 38292614 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021102.3 NP_066925.1 2 - 3 711 109
GenBank mRNA AB006534.1 - 2 - 3 588 109
AF027205.1 - 2 - 3 640 109
AK291532.1 - 2 - 3 526 109
BC001668.2 - 2 - 3 607 109
BC007705.2 - 2 - 3 555 109
BC011951.1 - 2 - 3 511 109
BC011955.1 - 2 - 3 483 109
BC012868.2 - 2 - 3 598 109
BT020115.1 - 2 - 3 276 109
BT020116.1 - 2 - 3 276 109
BT020117.1 - 2 - 3 276 109
CR407604.1 - 2 - 3 276 109
U78095.1 - 2 - 3 576 109

View Full Product Details