The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135052.3 | NP_001128524.1 | 6 - 7 | 1104 | 93 |
NM_001174168.2 | NP_001167639.1 | 6 - 7 | 969 | 93 | |
GenBank mRNA | BC011399.1 | - | 6 - 7 | 953 | 93 |
Z29630.1 | - | 6 - 7 | 997 | 93 |