Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC43A1
Entrez Gene ID
8501
Gene Name
solute carrier family 43 member 1
Gene Aliases
LAT3, PB39, POV1, R00504
Chromosome Location
Chr.11: 57484531 - 57515954 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198810.1 NP_001185739.1 5 - 6 586 58
NM_003627.5 NP_003618.1 5 - 6 757 58
XM_005274358.4 XP_005274415.1 6 - 7 897 58
XM_017018451.1 XP_016873940.1 6 - 7 785 58
GenBank mRNA AB103033.1 - 5 - 6 724 58
AF045584.1 - 5 - 6 544 58
AK027732.1 - 3 - 4 322 58
BC001639.2 - 5 - 6 586 58

View Full Product Details