Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TSC1
Entrez Gene ID
7248
Gene Name
tuberous sclerosis 1
Gene Aliases
LAM, TSC
Chromosome Location
Chr.9: 132891348 - 132945269 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000368.4 NP_000359.1 8 - 9 968 75
NM_001162426.1 NP_001155898.1 8 - 9 968 75
NM_001162427.1 NP_001155899.1 7 - 8 815 75
XM_005272211.1 XP_005272268.1 8 - 9 1013 75
XM_006717271.1 XP_006717334.1 8 - 9 948 75
XM_011518979.2 XP_011517281.1 8 - 9 962 75
XM_017015096.1 XP_016870585.1 9 - 10 1427 75
XM_017015097.1 XP_016870586.1 9 - 10 1398 75
XM_017015098.1 XP_016870587.1 8 - 9 1013 75
XM_017015099.1 XP_016870588.1 7 - 8 1106 75
XM_017015100.1 XP_016870589.1 7 - 8 1077 75
XM_017015101.1 XP_016870590.1 7 - 8 1107 75
GenBank mRNA AB190910.1 - 8 - 9 930 75
AF013168.1 - 8 - 9 955 75
AK297326.1 - 7 - 8 850 75
AK299654.1 - 6 - 7 649 75
AK303030.1 - 7 - 8 798 75
AK308412.1 - 8 - 9 951 75
BC047772.2 - 8 - 9 941 75
BC070032.1 - 8 - 9 969 75
BC108668.1 - 8 - 9 958 75
BC121000.2 - 8 - 9 925 75
KJ534953.1 - 6 - 7 734 75

View Full Product Details