Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 40396459-40396967 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRX
Entrez Gene ID
57716
Gene Name
periaxin
Gene Aliases
CMT4F
UniGene
Hs.205457
Gene Chromosome Location
Chr.19: 40393762-40414718 on Build GRCh38
Transcripts
NM_020956.2 NM_181882.2 XM_011527171.2 XM_017027046.1 XM_017027047.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3225591 c.1461G>A p.P487P Chr.19: 40396891-40396891 on Build GRCh38
3534065 c.1592C>T p.P531L Chr.19: 40396760-40396760 on Build GRCh38
7338122 c.1502A>T p.E501V Chr.19: 40396850-40396850 on Build GRCh38
4768881 c.1774G>C p.E592Q Chr.19: 40396578-40396578 on Build GRCh38
5493929 c.1657G>T p.E553* Chr.19: 40396695-40396695 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144305922 Chr.19: 40396806-40396806 on Build GRCh38
rs192607896 Chr.19: 40396842-40396842 on Build GRCh38
rs746715672 Chr.19: 40396497-40396497 on Build GRCh38
rs559655526 Chr.19: 40396510-40396510 on Build GRCh38
rs946852986 Chr.19: 40396606-40396606 on Build GRCh38
rs527537921 Chr.19: 40396546-40396546 on Build GRCh38
rs113515574 Chr.19: 40396601-40396601 on Build GRCh38
rs1000191235 Chr.19: 40396657-40396657 on Build GRCh38
rs548918678 Chr.19: 40396567-40396567 on Build GRCh38
rs374396283 Chr.19: 40396649-40396649 on Build GRCh38
rs537664679 Chr.19: 40396524-40396524 on Build GRCh38
rs972719763 Chr.19: 40396538-40396538 on Build GRCh38
rs879029685 Chr.19: 40396588-40396588 on Build GRCh38
rs767044807 Chr.19: 40396876-40396876 on Build GRCh38
rs201829675 Chr.19: 40396628-40396628 on Build GRCh38
rs61733448 Chr.19: 40396701-40396701 on Build GRCh38
rs761771873 Chr.19: 40396507-40396507 on Build GRCh38
rs761264097 Chr.19: 40396641-40396641 on Build GRCh38
rs148742209 Chr.19: 40396814-40396814 on Build GRCh38
rs567690884 Chr.19: 40396774-40396774 on Build GRCh38
rs754257849 Chr.19: 40396698-40396698 on Build GRCh38
rs878921895 Chr.19: 40396843-40396843 on Build GRCh38
rs147353593 Chr.19: 40396585-40396585 on Build GRCh38
rs753734869 Chr.19: 40396887-40396887 on Build GRCh38
rs879226255 Chr.19: 40396870-40396870 on Build GRCh38
rs772493394 Chr.19: 40396716-40396716 on Build GRCh38
rs757797949 Chr.19: 40396653-40396653 on Build GRCh38
rs762992156 Chr.19: 40396553-40396567 on Build GRCh38
rs898471400 Chr.19: 40396775-40396775 on Build GRCh38
rs756737014 Chr.19: 40396536-40396536 on Build GRCh38
rs1018810918 Chr.19: 40396871-40396871 on Build GRCh38
rs768022787 Chr.19: 40396534-40396534 on Build GRCh38
rs1001661757 Chr.19: 40396752-40396752 on Build GRCh38
rs201337455 Chr.19: 40396728-40396728 on Build GRCh38
rs747269937 Chr.19: 40396840-40396840 on Build GRCh38
rs759029769 Chr.19: 40396600-40396600 on Build GRCh38
rs750413570 Chr.19: 40396893-40396893 on Build GRCh38
rs752095452 Chr.19: 40396778-40396778 on Build GRCh38
rs778936443 Chr.19: 40396892-40396892 on Build GRCh38
rs780324179 Chr.19: 40396901-40396901 on Build GRCh38
rs878890753 Chr.19: 40396804-40396804 on Build GRCh38
rs781012349 Chr.19: 40396577-40396577 on Build GRCh38
rs754488421 Chr.19: 40396759-40396759 on Build GRCh38
rs146789340 Chr.19: 40396869-40396869 on Build GRCh38
rs767973929 Chr.19: 40396637-40396637 on Build GRCh38
rs567426170 Chr.19: 40396570-40396570 on Build GRCh38
rs377037656 Chr.19: 40396509-40396509 on Build GRCh38
rs1028086093 Chr.19: 40396486-40396486 on Build GRCh38
rs137865843 Chr.19: 40396675-40396675 on Build GRCh38
rs879133309 Chr.19: 40396822-40396822 on Build GRCh38
rs780959462 Chr.19: 40396667-40396667 on Build GRCh38
rs771684409 Chr.19: 40396489-40396489 on Build GRCh38
rs773222045 Chr.19: 40396515-40396515 on Build GRCh38
rs751136125 Chr.19: 40396756-40396756 on Build GRCh38
rs879254301 Chr.19: 40396694-40396694 on Build GRCh38
rs759927281 Chr.19: 40396527-40396527 on Build GRCh38
rs187786861 Chr.19: 40396813-40396813 on Build GRCh38
rs550446238 Chr.19: 40396784-40396784 on Build GRCh38
rs749156984 Chr.19: 40396799-40396799 on Build GRCh38
rs751816156 Chr.19: 40396525-40396525 on Build GRCh38
rs879923664 Chr.19: 40396937-40396937 on Build GRCh38
rs752314625 Chr.19: 40396883-40396883 on Build GRCh38
rs878945265 Chr.19: 40396864-40396864 on Build GRCh38
rs773206299 Chr.19: 40396593-40396593 on Build GRCh38
rs764542785 Chr.19: 40396731-40396731 on Build GRCh38
rs777104457 Chr.19: 40396852-40396852 on Build GRCh38
rs749264690 Chr.19: 40396599-40396599 on Build GRCh38
rs142723812 Chr.19: 40396561-40396561 on Build GRCh38
rs747179548 Chr.19: 40396911-40396911 on Build GRCh38
rs879110222 Chr.19: 40396792-40396792 on Build GRCh38
rs748484560 Chr.19: 40396940-40396940 on Build GRCh38
rs754681194 Chr.19: 40396666-40396666 on Build GRCh38
rs902117465 Chr.19: 40396874-40396874 on Build GRCh38
rs754382798 Chr.19: 40396650-40396650 on Build GRCh38
rs762424034 Chr.19: 40396736-40396736 on Build GRCh38
rs746077469 Chr.19: 40396715-40396715 on Build GRCh38
rs767098112 Chr.19: 40396767-40396767 on Build GRCh38
rs770161569 Chr.19: 40396945-40396945 on Build GRCh38
rs779516551 Chr.19: 40396559-40396559 on Build GRCh38
rs753226939 Chr.19: 40396535-40396535 on Build GRCh38
rs765791679 Chr.19: 40396739-40396739 on Build GRCh38
rs757931043 Chr.19: 40396555-40396555 on Build GRCh38
rs1043949955 Chr.19: 40396627-40396627 on Build GRCh38
rs879096800 Chr.19: 40396858-40396858 on Build GRCh38
rs1035002145 Chr.19: 40396805-40396805 on Build GRCh38
rs184025564 Chr.19: 40396636-40396636 on Build GRCh38
rs542511594 Chr.19: 40396594-40396594 on Build GRCh38
rs574899855 Chr.19: 40396516-40396516 on Build GRCh38
rs763786037 Chr.19: 40396885-40396885 on Build GRCh38
rs199629485 Chr.19: 40396692-40396692 on Build GRCh38
rs879234073 Chr.19: 40396783-40396783 on Build GRCh38
rs751183515 Chr.19: 40396663-40396663 on Build GRCh38
rs150582069 Chr.19: 40396706-40396706 on Build GRCh38
rs781271734 Chr.19: 40396933-40396933 on Build GRCh38
rs925714253 Chr.19: 40396924-40396924 on Build GRCh38
rs768402328 Chr.19: 40396569-40396569 on Build GRCh38
rs779133048 Chr.19: 40396809-40396809 on Build GRCh38
rs746698242 Chr.19: 40396562-40396562 on Build GRCh38
rs768453765 Chr.19: 40396498-40396498 on Build GRCh38
rs776504709 Chr.19: 40396506-40396506 on Build GRCh38
rs149715830 Chr.19: 40396778-40396778 on Build GRCh38
rs1055634958 Chr.19: 40396578-40396578 on Build GRCh38
rs779898963 Chr.19: 40396495-40396495 on Build GRCh38
rs764208864 Chr.19: 40396750-40396751 on Build GRCh38
rs139586219 Chr.19: 40396727-40396727 on Build GRCh38
rs879011078 Chr.19: 40396824-40396824 on Build GRCh38
rs200483634 Chr.19: 40396791-40396791 on Build GRCh38
rs546393524 Chr.19: 40396735-40396735 on Build GRCh38
rs368655227 Chr.19: 40396758-40396758 on Build GRCh38
rs759875870 Chr.19: 40396631-40396631 on Build GRCh38
rs778475989 Chr.19: 40396550-40396550 on Build GRCh38
rs370979792 Chr.19: 40396638-40396638 on Build GRCh38
rs757009052 Chr.19: 40396891-40396891 on Build GRCh38
rs950637477 Chr.19: 40396839-40396839 on Build GRCh38
rs768987878 Chr.19: 40396931-40396931 on Build GRCh38
rs369669118 Chr.19: 40396859-40396859 on Build GRCh38
rs758516996 Chr.19: 40396898-40396898 on Build GRCh38
rs1028012392 Chr.19: 40396723-40396723 on Build GRCh38
rs774393883 Chr.19: 40396626-40396626 on Build GRCh38
rs774541568 Chr.19: 40396714-40396714 on Build GRCh38
rs202113722 Chr.19: 40396705-40396705 on Build GRCh38
rs878911612 Chr.19: 40396780-40396780 on Build GRCh38
rs367946424 Chr.19: 40396873-40396873 on Build GRCh38
rs780375180 Chr.19: 40396823-40396823 on Build GRCh38
rs760423138 Chr.19: 40396884-40396884 on Build GRCh38
rs773718377 Chr.19: 40396872-40396872 on Build GRCh38

View Full Product Details