The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000861.3 | NP_000852.1 | 2 - 3 | 160 | 96 |
XM_011533652.1 | XP_011531954.1 | 3 - 4 | 448 | 96 | |
XM_017006283.1 | XP_016861772.1 | 1 - 2 | 231 | 96 | |
GenBank mRNA | BC060802.1 | - | 2 - 3 | 160 | 96 |