Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- AP1G2
- Entrez Gene ID
-
8906
- Gene Name
- adaptor related protein complex 1 gamma 2 subunit
- Gene Aliases
-
G2AD
- Chromosome Location
-
Chr.14: 23559563 - 23569056 on Build GRCh38
View Full Product Details