Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC9A3
Entrez Gene ID
6550
Gene Name
solute carrier family 9 member A3
Gene Aliases
DIAR8, NHE-3, NHE3
Chromosome Location
Chr.5: 471986 - 524434 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284351.2 NP_001271280.1 2 - 3 627 85
NM_004174.3 NP_004165.2 2 - 3 627 85
GenBank mRNA BC101669.1 - 2 - 3 523 85
U28043.1 - 2 - 3 525 85

View Full Product Details