The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166107.1 | NP_001159579.1 | 5 - 6 | 1152 | 94 |
NM_005518.3 | NP_005509.1 | 6 - 7 | 1278 | 94 | |
XM_011541313.1 | XP_011539615.1 | 5 - 6 | 1306 | 94 | |
GenBank mRNA | AK301594.1 | - | 5 - 6 | 935 | 94 |
AK302276.1 | - | 5 - 6 | 1121 | 94 | |
AK303777.1 | - | 5 - 6 | 1125 | 94 | |
AK313590.1 | - | 6 - 7 | 1237 | 94 | |
BC044217.1 | - | 6 - 7 | 1231 | 94 | |
CR456850.1 | - | 7 - 8 | 1190 | 94 | |
U12789.1 | - | 5 - 6 | 757 | 94 | |
X83618.1 | - | 6 - 7 | 1241 | 94 |