The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005811.3 | NP_005802.1 | 1 - 2 | 484 | 64 |
XM_006719194.3 | XP_006719257.1 | 1 - 2 | 486 | 64 | |
GenBank mRNA | AF028333.1 | - | 1 - 2 | 365 | 64 |
AF100907.1 | - | 1 - 2 | 485 | 64 |