Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SSR4
Entrez Gene ID
6748
Gene Name
signal sequence receptor subunit 4
Gene Aliases
CDG1Y, TRAPD
Chromosome Location
Chr.X: 153794175 - 153798512 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204526.1 NP_001191455.1 3 - 4 337 62
NM_001204527.1 NP_001191456.1 3 - 4 360 62
NM_006280.2 NP_006271.1 2 - 3 310 62
NR_037927.1 4 - 5 531 62
XM_011531186.1 XP_011529488.1 3 - 4 219 62
XM_011531187.1 XP_011529489.1 3 - 4 219 62
XM_017029756.1 XP_016885245.1 3 - 4 1108 62
XM_017029757.1 XP_016885246.1 3 - 4 843 62
GenBank mRNA AK057117.1 - 1 - 2 1893 62
AK290493.1 - 2 - 3 216 62
BC003371.1 - 2 - 3 190 62
BE791319.1 - 2 - 3 232 62
BG165852.1 - 3 - 4 344 62
BT007192.1 - 2 - 3 186 62
CD672615.1 - 3 - 4 402 62
X90583.1 - 2 - 3 216 62
Z69043.1 - 2 - 3 235 62

View Full Product Details