The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001078166.1 | NP_001071634.1 | 1 - 2 | 404 | 55 |
NM_006924.4 | NP_008855.1 | 1 - 2 | 404 | 55 | |
NR_034041.1 | 1 - 2 | 404 | 55 | ||
GenBank mRNA | AB062124.1 | - | 1 - 2 | 285 | 55 |
AB209558.1 | - | 1 - 2 | 291 | 55 | |
AK225711.1 | - | 1 - 2 | 282 | 55 | |
AK312781.1 | - | 1 - 2 | 301 | 55 | |
BC010264.1 | - | 1 - 2 | 230 | 55 | |
BC033785.1 | - | 1 - 2 | 290 | 55 | |
DA939911.1 | - | 1 - 2 | 404 | 55 | |
M69040.1 | - | 1 - 2 | 319 | 55 |