Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDK20
Entrez Gene ID
23552
Gene Name
cyclin dependent kinase 20
Gene Aliases
CCRK, CDCH, P42, PNQALRE
Chromosome Location
Chr.9: 87966441 - 87974780 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039803.2 NP_001034892.1 7 - 8 1181 87
NM_001170640.1 NP_001164111.1 5 - 6 994 87
NM_012119.4 NP_036251.2 6 - 7 1118 87
NM_178432.3 NP_848519.1 6 - 7 1157 87
XM_017014561.1 XP_016870050.1 7 - 8 1126 87
GenBank mRNA AF035013.1 - 6 - 7 1021 87
AF113130.1 - 4 - 5 2331 87
AK075325.1 - 6 - 7 910 87
AK294193.1 - 6 - 7 1079 87
AK298993.1 - 5 - 6 737 87
AY904367.1 - 5 - 6 660 87
BX647489.1 - 4 - 5 2349 87
KU178574.1 - 7 - 8 847 87
KU178575.1 - 6 - 7 784 87
KU178576.1 - 5 - 6 618 87
KU178577.1 - 4 - 5 538 87

View Full Product Details