The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012327.5 | NP_036459.1 | 5 - 6 | 816 | 96 |
NM_176787.4 | NP_789744.1 | 6 - 7 | 893 | 96 | |
XM_011525889.1 | XP_011524191.1 | 6 - 7 | 861 | 96 | |
XM_011525890.1 | XP_011524192.1 | 6 - 7 | 1058 | 96 | |
XM_011525891.1 | XP_011524193.1 | 6 - 7 | 776 | 96 | |
XM_011525892.1 | XP_011524194.1 | 5 - 6 | 798 | 96 | |
XM_011525893.1 | XP_011524195.1 | 5 - 6 | 749 | 96 | |
XM_011525894.1 | XP_011524196.1 | 5 - 6 | 660 | 96 | |
XM_011525895.1 | XP_011524197.1 | 4 - 5 | 658 | 96 | |
XM_011525896.1 | XP_011524198.1 | 6 - 7 | 861 | 96 | |
XM_011525898.1 | XP_011524200.1 | 6 - 7 | 861 | 96 | |
XM_017025685.1 | XP_016881174.1 | 6 - 7 | 861 | 96 | |
XM_017025686.1 | XP_016881175.1 | 3 - 4 | 451 | 96 | |
GenBank mRNA | AF109219.1 | - | 5 - 6 | 729 | 96 |
AK315134.1 | - | 6 - 7 | 861 | 96 | |
AL137607.1 | - | 2 - 3 | 128 | 96 | |
BC028363.2 | - | 6 - 7 | 830 | 96 |