The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014405.3 | NP_055220.1 | 2 - 3 | 353 | 85 |
GenBank mRNA | AF162692.1 | - | 2 - 3 | 322 | 85 |
AK315149.1 | - | 2 - 3 | 319 | 85 | |
BC034532.1 | - | 2 - 3 | 307 | 85 |