The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001167580.1 | NP_001161052.1 | 11 - 12 | 1832 | 90 |
NM_001323031.1 | NP_001309960.1 | 12 - 13 | 2365 | 90 | |
NM_001323032.1 | NP_001309961.1 | 12 - 13 | 2674 | 90 | |
NM_001323033.1 | NP_001309962.1 | 10 - 11 | 2341 | 90 | |
NM_001323034.1 | NP_001309963.1 | 11 - 12 | 2473 | 90 | |
NM_001323036.1 | NP_001309965.1 | 11 - 12 | 1651 | 90 | |
NM_001323037.1 | NP_001309966.1 | 13 - 14 | 2581 | 90 | |
NM_001323038.1 | NP_001309967.1 | 12 - 13 | 2493 | 90 | |
NM_001323039.1 | NP_001309968.1 | 12 - 13 | 2766 | 90 | |
NM_001323040.1 | NP_001309969.1 | 12 - 13 | 2012 | 90 | |
NM_014848.5 | NP_055663.1 | 13 - 14 | 2762 | 90 | |
XM_005254998.3 | XP_005255055.1 | 12 - 13 | 2427 | 90 | |
XM_017022761.1 | XP_016878250.1 | 12 - 13 | 2378 | 90 | |
XM_017022762.1 | XP_016878251.1 | 12 - 13 | 2513 | 90 | |
GenBank mRNA | AB018278.1 | - | 11 - 12 | 2035 | 90 |
AK294902.1 | - | 11 - 12 | 1834 | 90 | |
BC030011.2 | - | 13 - 14 | 2399 | 90 | |
CR457086.1 | - | 11 - 12 | 1871 | 90 |