The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015393.3 | NP_056208.2 | 3 - 4 | 1104 | 62 |
XM_011531833.1 | XP_011530135.1 | 4 - 5 | 1168 | 62 | |
GenBank mRNA | AK022311.1 | - | 3 - 4 | 1063 | 62 |
AL080121.1 | - | 3 - 4 | 1020 | 62 | |
AY358777.1 | - | 3 - 4 | 1049 | 62 | |
BC013294.2 | - | 3 - 4 | 1033 | 62 |