Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RRNAD1
Entrez Gene ID
51093
Gene Name
ribosomal RNA adenine dimethylase domain containing 1
Gene Aliases
C1orf66, CGI-41, METTL25B
Chromosome Location
Chr.1: 156728471 - 156736960 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142560.1 NP_001136032.1 1 - 2 744 67
NM_015997.3 NP_057081.3 1 - 2 744 67
XM_005245234.4 XP_005245291.1 1 - 2 529 67
XM_005245236.4 XP_005245293.1 1 - 2 529 67
XM_005245237.4 XP_005245294.1 1 - 2 529 67
XM_017001405.1 XP_016856894.1 1 - 2 529 67
XM_017001406.1 XP_016856895.1 1 - 2 529 67
GenBank mRNA AF151800.1 - 1 - 2 334 67
AK000412.1 - 2 - 3 560 67
BC011382.1 - 1 - 2 729 67

View Full Product Details