Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NT5DC3
Entrez Gene ID
51559
Gene Name
5'-nucleotidase domain containing 3
Gene Aliases
TU12B1-TY, TU12B1TY
Chromosome Location
Chr.12: 103746315 - 103841238 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031701.2 NP_001026871.1 2 - 3 438 76
XM_017019454.1 XP_016874943.1 2 - 3 479 76
GenBank mRNA AB032773.1 - 2 - 3 248 76
BC052293.1 - 2 - 3 438 76

View Full Product Details