Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
A4GALT
Entrez Gene ID
53947
Gene Name
alpha 1,4-galactosyltransferase
Gene Aliases
A14GALT, A4GALT1, Gb3S, P(k), P1, P1PK, PK
Chromosome Location
Chr.22: 42692115 - 42721301 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318038.1 NP_001304967.1 2 - 3 276 74
NM_017436.5 NP_059132.1 2 - 3 253 74
XM_006724265.3 XP_006724328.1 2 - 3 499 74
XM_006724266.3 XP_006724329.1 2 - 3 233 74
GenBank mRNA AB037883.1 - 1 - 2 85 74
AK312927.1 - 2 - 3 257 74
BC017068.2 - 2 - 3 172 74
CB150578.1 - 2 - 3 214 74
HY140390.1 - 2 - 3 254 74

View Full Product Details