Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRPM6
Entrez Gene ID
140803
Gene Name
transient receptor potential cation channel subfamily M member 6
Gene Aliases
CHAK2, HMGX, HOMG, HOMG1, HSH
Chromosome Location
Chr.9: 74722495 - 74888094 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177310.1 NP_001170781.1 33 - 34 5262 72
NM_001177311.1 NP_001170782.1 33 - 34 5289 72
NM_017662.4 NP_060132.3 33 - 34 5443 72
XM_011518245.1 XP_011516547.1 32 - 33 5350 72
XM_011518246.1 XP_011516548.1 33 - 34 5443 72
XM_011518247.1 XP_011516549.1 32 - 33 5314 72
XM_011518248.1 XP_011516550.1 32 - 33 5302 72
XM_011518249.1 XP_011516551.1 31 - 32 5209 72
XM_011518250.1 XP_011516552.1 31 - 32 5167 72
XM_011518251.2 XP_011516553.1 28 - 29 4694 72
XM_017014287.1 XP_016869776.1 30 - 31 5080 72
XM_017014288.1 XP_016869777.1 29 - 30 4933 72
GenBank mRNA AF350881.2 - 33 - 34 5443 72
AF448232.1 - 33 - 34 5205 72
AK000094.1 - 10 - 11 2169 72
AK225207.1 - 10 - 11 2169 72
AY333282.1 - 33 - 34 5268 72
AY333283.1 - 33 - 34 5262 72
AY333284.1 - 33 - 34 5289 72
AY333285.1 - 33 - 34 5268 72
AY333286.1 - 20 - 21 2121 72
AY333287.1 - 17 - 18 1767 72

View Full Product Details