The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017729.3 | NP_060199.3 | 9 - 10 | 1111 | 99 |
NM_133180.2 | NP_573441.2 | 14 - 15 | 1494 | 99 | |
GenBank mRNA | AF370395.1 | - | 14 - 15 | 1446 | 99 |
AK000265.1 | - | 9 - 10 | 1111 | 99 | |
AK075098.1 | - | 14 - 15 | 1445 | 99 | |
AK296732.1 | - | 12 - 13 | 1344 | 99 | |
AY074928.1 | - | 13 - 14 | 1447 | 99 | |
BC015763.1 | - | 9 - 10 | 1080 | 99 |