Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HAUS2
Entrez Gene ID
55142
Gene Name
HAUS augmin like complex subunit 2
Gene Aliases
C15orf25, CEP27, HsT17025
Chromosome Location
Chr.15: 42548813 - 42569994 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001323629.1 NP_001310558.1 1 - 2 152 120
NM_001323630.1 NP_001310559.1 1 - 2 152 120
NM_001323631.1 NP_001310560.1 1 - 2 152 120
NM_001323632.1 NP_001310561.1 1 - 2 152 120
NM_018097.2 NP_060567.1 1 - 2 152 120
XM_017022396.1 XP_016877885.1 1 - 2 137 120
GenBank mRNA AK001322.1 - 1 - 2 110 120
BC010903.1 - 1 - 2 131 120
DB450370.1 - 1 - 2 152 120

View Full Product Details