The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018941.3 | NP_061764.2 | 2 - 3 | 850 | 71 |
XM_005266021.3 | XP_005266078.1 | 2 - 3 | 794 | 71 | |
XM_005266022.1 | XP_005266079.1 | 2 - 3 | 973 | 71 | |
XM_005266023.1 | XP_005266080.1 | 2 - 3 | 691 | 71 | |
XM_011534745.1 | XP_011533047.1 | 2 - 3 | 795 | 71 | |
XM_011534746.2 | XP_011533048.1 | 2 - 3 | 692 | 71 | |
GenBank mRNA | AF123757.1 | - | 2 - 3 | 779 | 71 |
AF123758.1 | - | 2 - 3 | 779 | 71 | |
AF123759.1 | - | 2 - 3 | 779 | 71 | |
AF123760.1 | - | 2 - 3 | 779 | 71 | |
AF123761.1 | - | 2 - 3 | 779 | 71 | |
AK312486.1 | - | 2 - 3 | 793 | 71 | |
BC007725.2 | - | 2 - 3 | 704 | 71 | |
BT007049.1 | - | 1 - 2 | 545 | 71 |