The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145076.2 | NP_001138548.1 | 21 - 22 | 2575 | 90 |
NM_019063.4 | NP_061936.2 | 22 - 23 | 2749 | 90 | |
XM_005264267.1 | XP_005264324.1 | 23 - 24 | 2762 | 90 | |
XM_006711991.1 | XP_006712054.1 | 22 - 23 | 2588 | 90 | |
XM_006711992.2 | XP_006712055.1 | 24 - 25 | 2883 | 90 | |
GenBank mRNA | AF177377.2 | - | 22 - 23 | 2711 | 90 |
AK001804.1 | - | - | 175 | 90 | |
AK056880.1 | - | 11 - 12 | 1251 | 90 | |
AK295778.1 | - | 7 - 8 | 787 | 90 | |
AK314702.1 | - | 22 - 23 | 2723 | 90 | |
AL832392.1 | - | 8 - 9 | 902 | 90 | |
BC023522.2 | - | 1 - 2 | 139 | 90 | |
BC140845.1 | - | 22 - 23 | 2672 | 90 | |
BC146799.1 | - | 21 - 22 | 2549 | 90 | |
BX647731.1 | - | 13 - 14 | 1812 | 90 |