The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206671.2 | NP_001193600.1 | 5 - 6 | 731 | 83 |
NM_024557.4 | NP_078833.3 | 5 - 6 | 728 | 83 | |
NR_045405.1 | 6 - 7 | 780 | 83 | ||
XM_006718317.3 | XP_006718380.1 | 5 - 6 | 860 | 83 | |
XM_006718318.3 | XP_006718381.1 | 5 - 6 | 773 | 83 | |
XM_017018287.1 | XP_016873776.1 | 5 - 6 | 860 | 83 | |
XM_017018288.1 | XP_016873777.1 | 6 - 7 | 857 | 83 | |
XM_017018289.1 | XP_016873778.1 | 6 - 7 | 804 | 83 | |
GenBank mRNA | AK021670.1 | - | 4 - 5 | 752 | 83 |
AK293933.1 | - | 5 - 6 | 778 | 83 | |
AK296505.1 | - | 6 - 7 | 745 | 83 | |
AK315379.1 | - | 5 - 6 | 687 | 83 | |
AY326435.1 | - | 5 - 6 | 677 | 83 | |
BC022455.1 | - | 5 - 6 | 710 | 83 | |
KU178773.1 | - | 5 - 6 | 674 | 83 | |
KU178774.1 | - | 4 - 5 | 580 | 83 |