The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301039.1 | NP_001287968.1 | 2 - 3 | 198 | 73 |
NM_001301040.1 | NP_001287969.1 | 2 - 3 | 198 | 73 | |
NM_001301042.1 | NP_001287971.1 | 1 - 2 | 68 | 73 | |
NM_024579.3 | NP_078855.2 | 1 - 2 | 87 | 73 | |
GenBank mRNA | AA095731.1 | - | 1 - 2 | 55 | 73 |
AI342055.1 | - | 4 - 5 | 73 | ||
AK026874.1 | - | 1 - 2 | 72 | 73 | |
AX770997.1 | - | 1 - 2 | 85 | 73 | |
AX770998.1 | - | 2 - 3 | 112 | 73 | |
AX770999.1 | - | 1 - 2 | 87 | 73 | |
BC017761.1 | - | 1 - 2 | 72 | 73 |