Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ALG13
Entrez Gene ID
79868
Gene Name
ALG13, UDP-N-acetylglucosaminyltransferase subunit
Gene Aliases
CDG1S, CXorf45, EIEE36, GLT28D1, MDS031, TDRD13, YGL047W
Chromosome Location
Chr.X: 111680741 - 111764164 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099922.2 NP_001093392.1 19 - 20 2472 74
NM_001257230.1 NP_001244159.1 19 - 20 2368 74
NM_001257231.1 NP_001244160.1 19 - 20 2486 74
NM_001257234.1 NP_001244163.1 19 - 20 2441 74
NM_001257237.1 NP_001244166.1 19 - 20 2545 74
NM_001324292.1 NP_001311221.1 19 - 20 2472 74
NM_001324293.1 NP_001311222.1 17 - 18 2371 74
XM_006724693.2 XP_006724756.1 19 - 20 2845 74
XM_006724697.3 XP_006724760.2 19 - 20 2745 74
XM_006724698.3 XP_006724761.2 19 - 20 2870 74
XM_011531028.2 XP_011529330.1 19 - 20 2846 74
XM_011531030.1 XP_011529332.1 19 - 20 2844 74
XM_011531031.1 XP_011529333.1 19 - 20 2545 74
XM_011531032.1 XP_011529334.1 19 - 20 2351 74
XM_011531033.2 XP_011529335.1 19 - 20 2391 74
XM_011531034.2 XP_011529336.1 17 - 18 2085 74
XM_011531037.1 XP_011529339.1 9 - 10 1102 74
XM_017029846.1 XP_016885335.1 19 - 20 2823 74
XM_017029847.1 XP_016885336.1 19 - 20 2846 74
XM_017029848.1 XP_016885337.1 19 - 20 2820 74
XM_017029849.1 XP_016885338.1 19 - 20 2845 74
XM_017029850.1 XP_016885339.1 19 - 20 2844 74
XM_017029851.1 XP_016885340.1 20 - 21 2753 74
XM_017029852.1 XP_016885341.1 12 - 13 1418 74
XM_017029853.1 XP_016885342.1 15 - 16 1660 74
XM_017029854.1 XP_016885343.1 6 - 7 1182 74
XM_017029855.1 XP_016885344.1 12 - 13 2110 74
GenBank mRNA AK026671.1 - 4 - 5 601 74
AK223154.1 - 4 - 5 601 74
AK300394.1 - 19 - 20 2310 74
AK302729.1 - 19 - 20 2413 74
AK302890.1 - 19 - 20 2389 74
AK304712.1 - 19 - 20 2362 74
AK316306.1 - 19 - 20 2414 74
AK316522.1 - 19 - 20 2489 74
BC117377.1 - 4 - 5 599 74
BC117379.1 - 4 - 5 599 74

View Full Product Details