The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001288968.1 | NP_001275897.1 | 11 - 12 | 1395 | 77 |
NM_001288969.1 | NP_001275898.1 | 10 - 11 | 1281 | 77 | |
NM_001288970.1 | NP_001275899.1 | 9 - 10 | 1191 | 77 | |
NM_001288971.1 | NP_001275900.1 | 8 - 9 | 995 | 77 | |
NM_001288972.1 | NP_001275901.1 | 10 - 11 | 1237 | 77 | |
NM_025149.5 | NP_079425.3 | 10 - 11 | 1320 | 77 | |
XM_006722110.3 | XP_006722173.1 | 10 - 11 | 1260 | 77 | |
GenBank mRNA | AK024573.1 | - | 10 - 11 | 1227 | 77 |
AK225044.1 | - | 10 - 11 | 1227 | 77 | |
AK225561.1 | - | 10 - 11 | 1226 | 77 | |
AK294205.1 | - | 11 - 12 | 1337 | 77 | |
AK295258.1 | - | 10 - 11 | 1189 | 77 | |
AK300145.1 | - | 9 - 10 | 1099 | 77 | |
AK300625.1 | - | 8 - 9 | 995 | 77 | |
AY358660.1 | - | 10 - 11 | 1256 | 77 | |
BC012053.1 | - | 10 - 11 | 1233 | 77 | |
BC014123.2 | - | 10 - 11 | 1240 | 77 |