The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030974.3 | NP_112236.3 | 2 - 3 | 859 | 75 |
NR_038270.1 | 2 - 3 | 857 | 75 | ||
XM_017013887.1 | XP_016869376.1 | 2 - 3 | 887 | 75 | |
XM_017013888.1 | XP_016869377.1 | 2 - 3 | 887 | 75 | |
GenBank mRNA | AB052764.1 | - | 2 - 3 | 857 | 75 |
AK075518.1 | - | 2 - 3 | 814 | 75 | |
AL136816.1 | - | 2 - 3 | 583 | 75 | |
BC025244.1 | - | 2 - 3 | 859 | 75 | |
BC034028.1 | - | 2 - 3 | 388 | 75 | |
BE877614.1 | - | 2 - 3 | 324 | 75 | |
FJ655995.1 | - | 2 - 3 | 645 | 75 |