The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271985.1 | NP_001258914.1 | 2 - 3 | 612 | 132 |
NM_031307.3 | NP_112597.3 | 3 - 4 | 1036 | 132 | |
XM_005271687.3 | XP_005271744.1 | 2 - 3 | 1165 | 132 | |
GenBank mRNA | AF325689.1 | - | 2 - 3 | 968 | 132 |
AK055702.1 | - | 3 - 4 | 1013 | 132 | |
AK314251.1 | - | 3 - 4 | 1034 | 132 | |
BC004822.1 | - | 3 - 4 | 1025 | 132 | |
BC013427.2 | - | 1 - 2 | 256 | 132 | |
CR982568.1 | - | 2 - 3 | 588 | 132 |