Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BMP6
Entrez Gene ID
654
Gene Name
bone morphogenetic protein 6
Gene Aliases
VGR, VGR1
Chromosome Location
Chr.6: 7726778 - 7881728 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001718.4 NP_001709.1 2 - 3 1036 128
GenBank mRNA AK300628.1 - 3 - 4 996 128
BC096084.2 - 3 - 4 604 128
BC096085.2 - 3 - 4 604 128
BC099624.1 - 3 - 4 604 128
BC099625.1 - 3 - 4 768 128
BC111763.1 - 3 - 4 768 128
BC112352.1 - 3 - 4 768 128
BC113018.1 - 3 - 4 663 128
BC113019.1 - 3 - 4 660 128
BC128031.1 - 3 - 4 560 128
M60315.1 - 2 - 3 1017 128

View Full Product Details