The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001740.4 | NP_001731.2 | 9 - 10 | 713 | 67 |
NR_027910.2 | 8 - 9 | 673 | 67 | ||
GenBank mRNA | AK222495.1 | - | 9 - 10 | 703 | 67 |
AK222648.1 | - | 9 - 10 | 703 | 67 | |
AK291096.1 | - | 9 - 10 | 703 | 67 | |
BC015484.2 | - | 9 - 10 | 688 | 67 | |
X56667.1 | - | 9 - 10 | 676 | 67 |