Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LYL1
Entrez Gene ID
4066
Gene Name
LYL1, basic helix-loop-helix family member
Gene Aliases
bHLHa18
Chromosome Location
Chr.19: 13099028 - 13103160 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005583.4 NP_005574.2 3 - 4 1085 65
XM_005259912.3 XP_005259969.1 3 - 4 923 65
XM_017026816.1 XP_016882305.1 4 - 5 1785 65
XM_017026817.1 XP_016882306.1 4 - 5 1638 65
GenBank mRNA BC002796.2 - 3 - 4 792 65
M22637.1 - 3 - 4 650 65

View Full Product Details