Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PCDHGB1
Entrez Gene ID
56104
Gene Name
protocadherin gamma subfamily B, 1
Gene Aliases
PCDH-GAMMA-B1
Chromosome Location
Chr.5: 141350261 - 141512979 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018922.2 NP_061745.1 1 - 2 2407 63
GenBank mRNA AF152330.1 - 1 - 2 2407 63

View Full Product Details