Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PCDHGB2
Entrez Gene ID
56103
Gene Name
protocadherin gamma subfamily B, 2
Gene Aliases
PCDH-GAMMA-B2
Chromosome Location
Chr.5: 141360136 - 141512979 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018923.2 NP_061746.1 1 - 2 2426 62
GenBank mRNA AF152331.1 - 1 - 2 2426 62

View Full Product Details