Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 225296-225804 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SDHA
Entrez Gene ID
6389
Gene Name
succinate dehydrogenase complex flavoprotein subunit A
Gene Aliases
CMD1GG, FP, PGL5, SDH1, SDH2, SDHF
UniGene
Hs.440475
Gene Chromosome Location
Chr.5: 218223-264816 on Build GRCh38
Transcripts
NM_001294332.1 NM_004168.3 XM_005248331.3 XM_011514072.1 XM_011514073.1 XM_017009685.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6374247 c.407A>T p.Q136L Chr.5: 225513-225513 on Build GRCh38
7423663 c.424A>T p.M142L Chr.5: 225530-225530 on Build GRCh38
5868371 c.379G>A p.V127M Chr.5: 225485-225485 on Build GRCh38
6404042 c.359G>T p.R120M Chr.5: 225465-225465 on Build GRCh38
6420060 c.445G>A p.A149T Chr.5: 225551-225551 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762380125 Chr.5: 225403-225403 on Build GRCh38
rs772169255 Chr.5: 225379-225379 on Build GRCh38
rs191318615 Chr.5: 225378-225378 on Build GRCh38
rs754393360 Chr.5: 225478-225478 on Build GRCh38
rs56342538 Chr.5: 225681-225681 on Build GRCh38
rs763191656 Chr.5: 225546-225546 on Build GRCh38
rs375645919 Chr.5: 225442-225442 on Build GRCh38
rs756039268 Chr.5: 225484-225484 on Build GRCh38
rs373761366 Chr.5: 225373-225373 on Build GRCh38
rs752187837 Chr.5: 225553-225553 on Build GRCh38
rs377528979 Chr.5: 225377-225377 on Build GRCh38
rs530954561 Chr.5: 225343-225343 on Build GRCh38
rs200565489 Chr.5: 225571-225571 on Build GRCh38
rs749893972 Chr.5: 225610-225610 on Build GRCh38
rs12520059 Chr.5: 225653-225653 on Build GRCh38
rs750806202 Chr.5: 225406-225406 on Build GRCh38
rs987288514 Chr.5: 225459-225459 on Build GRCh38
rs11557099 Chr.5: 225490-225490 on Build GRCh38
rs776848209 Chr.5: 225530-225530 on Build GRCh38
rs933414586 Chr.5: 225446-225446 on Build GRCh38
rs528774217 Chr.5: 225702-225702 on Build GRCh38
rs541934209 Chr.5: 225648-225648 on Build GRCh38
rs1002981208 Chr.5: 225350-225350 on Build GRCh38
rs35464369 Chr.5: 225594-225594 on Build GRCh38
rs763490338 Chr.5: 225583-225583 on Build GRCh38
rs765884036 Chr.5: 225437-225437 on Build GRCh38
rs748121363 Chr.5: 225483-225483 on Build GRCh38
rs751114575 Chr.5: 225440-225440 on Build GRCh38
rs765766175 Chr.5: 225404-225404 on Build GRCh38
rs764728460 Chr.5: 225397-225397 on Build GRCh38
rs759266253 Chr.5: 225525-225525 on Build GRCh38
rs773634017 Chr.5: 225542-225542 on Build GRCh38
rs368286780 Chr.5: 225451-225451 on Build GRCh38
rs190477316 Chr.5: 225514-225514 on Build GRCh38
rs777600956 Chr.5: 225485-225485 on Build GRCh38
rs1023347173 Chr.5: 225341-225341 on Build GRCh38
rs748976493 Chr.5: 225496-225496 on Build GRCh38
rs772343171 Chr.5: 225510-225510 on Build GRCh38
rs748175384 Chr.5: 225578-225578 on Build GRCh38
rs199805178 Chr.5: 225447-225447 on Build GRCh38
rs786205209 Chr.5: 225434-225434 on Build GRCh38
rs193283468 Chr.5: 225582-225582 on Build GRCh38
rs369382811 Chr.5: 225362-225362 on Build GRCh38
rs954767622 Chr.5: 225639-225639 on Build GRCh38
rs748550489 Chr.5: 225772-225772 on Build GRCh38
rs778737664 Chr.5: 225560-225560 on Build GRCh38
rs367618662 Chr.5: 225550-225550 on Build GRCh38
rs780005840 Chr.5: 225668-225668 on Build GRCh38
rs201972549 Chr.5: 225412-225412 on Build GRCh38
rs574620269 Chr.5: 225646-225646 on Build GRCh38
rs963377808 Chr.5: 225381-225381 on Build GRCh38
rs935241830 Chr.5: 225477-225477 on Build GRCh38
rs747232441 Chr.5: 225526-225526 on Build GRCh38
rs768897373 Chr.5: 225529-225529 on Build GRCh38
rs876660259 Chr.5: 225513-225513 on Build GRCh38
rs767097738 Chr.5: 225603-225603 on Build GRCh38
rs201453889 Chr.5: 225547-225547 on Build GRCh38
rs61492827 Chr.5: 225659-225659 on Build GRCh38
rs923803277 Chr.5: 225460-225460 on Build GRCh38
rs185113945 Chr.5: 225758-225758 on Build GRCh38
rs575617625 Chr.5: 225551-225551 on Build GRCh38
rs779223360 Chr.5: 225711-225711 on Build GRCh38
rs375576259 Chr.5: 225548-225548 on Build GRCh38
rs771236880 Chr.5: 225592-225592 on Build GRCh38
rs386684228 Chr.5: 225658-225659 on Build GRCh38
rs781436294 Chr.5: 225572-225572 on Build GRCh38
rs770457461 Chr.5: 225580-225580 on Build GRCh38
rs149101259 Chr.5: 225624-225624 on Build GRCh38
rs780654623 Chr.5: 225479-225479 on Build GRCh38
rs542980860 Chr.5: 225554-225554 on Build GRCh38
rs758086385 Chr.5: 225429-225429 on Build GRCh38
rs906281170 Chr.5: 225535-225535 on Build GRCh38
rs775651866 Chr.5: 225391-225391 on Build GRCh38
rs371735891 Chr.5: 225568-225568 on Build GRCh38
rs760604346 Chr.5: 225395-225395 on Build GRCh38
rs138917116 Chr.5: 225559-225559 on Build GRCh38
rs185555941 Chr.5: 225400-225400 on Build GRCh38
rs755246495 Chr.5: 225703-225703 on Build GRCh38
rs761345594 Chr.5: 225368-225369 on Build GRCh38
rs200675907 Chr.5: 225534-225534 on Build GRCh38
rs1046766708 Chr.5: 225753-225753 on Build GRCh38
rs1012325962 Chr.5: 225337-225337 on Build GRCh38
rs753947707 Chr.5: 225337-225337 on Build GRCh38
rs200342815 Chr.5: 225645-225645 on Build GRCh38
rs765784494 Chr.5: 225539-225539 on Build GRCh38
rs939024363 Chr.5: 225707-225707 on Build GRCh38
rs752206435 Chr.5: 225608-225608 on Build GRCh38
rs143148642 Chr.5: 225557-225557 on Build GRCh38
rs115037335 Chr.5: 225658-225658 on Build GRCh38
rs764993861 Chr.5: 225499-225502 on Build GRCh38

View Full Product Details