Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COL27A1
Entrez Gene ID
85301
Gene Name
collagen type XXVII alpha 1
Gene Aliases
STLS
Chromosome Location
Chr.9: 114154271 - 114312516 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032888.3 NP_116277.2 59 - 60 5624 62
XM_006717308.2 XP_006717371.1 58 - 59 5638 62
XM_006717310.3 XP_006717373.1 57 - 58 3437 62
XM_011519138.2 XP_011517440.1 59 - 60 5256 62
XM_011519140.1 XP_011517442.1 58 - 59 5638 62
XM_011519145.2 XP_011517447.1 46 - 47 2866 62
GenBank mRNA AB058773.1 - 56 - 57 4111 62
AK021957.1 - 1 - 2 369 62
AY149237.1 - 59 - 60 5218 62

View Full Product Details