Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
YIPF4
Entrez Gene ID
84272
Gene Name
Yip1 domain family member 4
Gene Aliases
FinGER4, Nbla11189
Chromosome Location
Chr.2: 32277889 - 32307929 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032312.3 NP_115688.1 3 - 4 670 118
XM_005264599.3 XP_005264656.1 3 - 4 664 118
GenBank mRNA AB072898.1 - 3 - 4 617 118
AK098486.1 - 3 - 4 649 118
BC004875.2 - 3 - 4 597 118

View Full Product Details