The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032588.3 | NP_115977.2 | 8 - 9 | 1195 | 95 |
GenBank mRNA | AF353673.1 | - | 8 - 9 | 1174 | 95 |
AJ276484.1 | - | 8 - 9 | 1178 | 95 | |
AK056942.1 | - | 8 - 9 | 1191 | 95 | |
AK297820.1 | - | 8 - 9 | 1107 | 95 | |
BC080529.1 | - | 8 - 9 | 1173 | 95 |