The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001126328.2 | NP_001119800.1 | 6 - 7 | 1633 | 105 |
NM_032622.2 | NP_116011.2 | 5 - 6 | 1364 | 105 | |
XM_005265785.4 | XP_005265842.1 | 6 - 7 | 1791 | 105 | |
XM_017008776.1 | XP_016864265.1 | 7 - 8 | 1852 | 105 | |
GenBank mRNA | AF237782.1 | - | 5 - 6 | 1295 | 105 |
AK056823.1 | - | 6 - 7 | 1606 | 105 | |
AK294611.1 | - | 6 - 7 | 1162 | 105 | |
AY358326.1 | - | 5 - 6 | 1275 | 105 | |
BC022983.1 | - | 6 - 7 | 1509 | 105 | |
BC034737.1 | - | 5 - 6 | 1284 | 105 |