Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYTL2
Entrez Gene ID
54843
Gene Name
synaptotagmin like 2
Gene Aliases
CHR11SYT, EXO4, PPP1R151, SGA72M, SLP2, SLP2A
Chromosome Location
Chr.11: 85694221 - 85811182 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001162951.2 NP_001156423.1 2 - 3 595 86
NM_001162953.2 NP_001156425.1 2 - 3 595 86
NM_001289608.1 NP_001276537.1 2 - 3 356 86
NM_032943.4 NP_116561.1 3 - 4 846 86
XM_005274057.1 XP_005274114.1 2 - 3 367 86
XM_005274060.1 XP_005274117.1 2 - 3 367 86
XM_005274061.1 XP_005274118.1 2 - 3 367 86
XM_005274063.1 XP_005274120.1 2 - 3 367 86
XM_005274066.1 XP_005274123.1 2 - 3 367 86
XM_011545105.1 XP_011543407.1 2 - 3 367 86
XM_011545106.1 XP_011543408.1 2 - 3 367 86
XM_011545108.1 XP_011543410.1 2 - 3 367 86
XM_011545109.2 XP_011543411.1 2 - 3 352 86
XM_011545110.1 XP_011543412.1 2 - 3 367 86
XM_011545112.1 XP_011543414.1 2 - 3 367 86
XM_017017933.1 XP_016873422.1 2 - 3 367 86
XM_017017935.1 XP_016873424.1 2 - 3 367 86
XM_017017938.1 XP_016873427.1 2 - 3 349 86
GenBank mRNA AB046817.1 - 3 - 4 857 86
AK000170.1 - 2 - 3 271 86
AK296224.1 - 2 - 3 338 86
BC136450.1 - 2 - 3 527 86
BC144114.1 - 2 - 3 527 86
BC144115.1 - 2 - 3 527 86
BQ308382.1 - 2 - 3 85
DA449325.1 - 1 - 2 132 86
EU035829.1 - 2 - 3 251 86

View Full Product Details