Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- SYTL2
- Entrez Gene ID
-
54843
- Gene Name
- synaptotagmin like 2
- Gene Aliases
-
CHR11SYT, EXO4, PPP1R151, SGA72M, SLP2, SLP2A
- Chromosome Location
-
Chr.11: 85694221 - 85811182 on Build GRCh38
View Full Product Details