The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256430.1 | NP_001243359.1 | 6 - 7 | 2825 | 60 |
NM_033104.3 | NP_149095.2 | 5 - 6 | 2721 | 60 | |
XM_006720279.1 | XP_006720342.1 | 4 - 5 | 2610 | 60 | |
XM_011537240.1 | XP_011535542.1 | 5 - 6 | 2781 | 60 | |
XM_011537241.1 | XP_011535543.1 | 5 - 6 | 2781 | 60 | |
XM_011537242.1 | XP_011535544.1 | 5 - 6 | 2781 | 60 | |
GenBank mRNA | AB208948.1 | - | 6 - 7 | 2825 | 60 |
AF255309.1 | - | 1 - 2 | 1920 | 60 | |
AF380833.1 | - | 4 - 5 | 2455 | 60 | |
AF449430.1 | - | 4 - 5 | 2613 | 60 | |
AK094799.1 | - | 1 - 2 | 1025 | 60 | |
BC069359.1 | - | 4 - 5 | 2593 | 60 | |
BC069389.1 | - | 4 - 5 | 2587 | 60 | |
BC069502.1 | - | 4 - 5 | 2593 | 60 | |
BC069587.1 | - | 4 - 5 | 2594 | 60 | |
BC117493.1 | - | 4 - 5 | 2523 | 60 | |
BC117495.1 | - | 4 - 5 | 2523 | 60 |